Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178258.5 | NP_839989.2 | 1 - 2 | 68 | 97 | |
GenBank mRNA | AF493604.1 | - | 1 - 2 | 56 | 97 | |
AJ555484.1 | - | 1 - 2 | 62 | 97 | ||
AK040641.1 | - | 1 - 2 | 77 | 97 | ||
AK138913.1 | - | 1 - 2 | 69 | 97 | ||
AK139024.1 | - | 1 - 2 | 99 | 97 | ||
BC124998.1 | - | 1 - 2 | 58 | 40044741 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
