Assay Details
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 11 - 12 | 1152 | 92 | |
GenBank mRNA | AK044772.1 | - | 11 - 12 | 1148 | 92 | |
AK082408.1 | - | 12 - 13 | 1254 | 92 | ||
AK163712.1 | - | 11 - 12 | 1156 | 92 | ||
BC132485.1 | - | 11 - 12 | 1127 | 40130762 | 92 | |
BC144986.1 | - | 12 - 13 | 1228 | 92 | ||
BC144987.1 | - | 11 - 12 | 1203 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
