Assay Details
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 16 - 17 | 1658 | 87 | |
GenBank mRNA | AK044772.1 | - | 16 - 17 | 1654 | 87 | |
AK046875.1 | - | 1 - 2 | 168 | 87 | ||
AK082408.1 | - | 17 - 18 | 1760 | 87 | ||
BC132485.1 | - | 16 - 17 | 1633 | 40130762 | 87 | |
BC144986.1 | - | 17 - 18 | 1734 | 87 | ||
BC144987.1 | - | 16 - 17 | 1709 | 87 | ||
BC144989.1 | - | 16 - 17 | 1632 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
