Assay Details
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171000.2 | NP_001164471.2 | 6 - 7 | 1013 | 85 | |
NM_001171001.1 | NP_001164472.1 | 6 - 7 | 733 | 85 | ||
NM_021414.6 | NP_067389.5 | 6 - 7 | 1016 | 85 | ||
GenBank mRNA | AK014539.1 | - | 6 - 7 | 964 | 85 | |
AK053527.1 | - | 6 - 7 | 697 | 85 | ||
AK122382.1 | - | 4 - 5 | 719 | 85 | ||
BC027436.1 | - | 6 - 7 | 928 | 85 | ||
BC059255.1 | - | 2 - 3 | 187 | 85 | ||
BC079660.1 | - | 6 - 7 | 926 | 6853550 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
