Assay Details
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143776.1 | NP_001137248.1 | 3 - 4 | 428 | 72 | |
NM_001143777.1 | NP_001137249.1 | 4 - 5 | 532 | 72 | ||
NM_024244.4 | NP_077206.3 | 3 - 4 | 428 | 72 | ||
GenBank mRNA | AK004797.1 | - | 3 - 4 | 412 | 72 | |
AK021133.1 | - | 4 - 5 | 371 | 72 | ||
AK032053.1 | - | 4 - 5 | 344 | 72 | ||
AK036299.1 | - | 4 - 5 | 495 | 72 | ||
AK039513.1 | - | 4 - 5 | 496 | 72 | ||
AK041147.1 | - | 3 - 4 | 429 | 72 | ||
AK044049.1 | - | 4 - 5 | 455 | 72 | ||
AK081939.1 | - | 4 - 5 | 533 | 72 | ||
AK082202.1 | - | 3 - 4 | 424 | 72 | ||
AK220421.1 | - | 4 - 5 | 368 | 72 | ||
BC129839.1 | - | 3 - 4 | 384 | 40060810 | 72 | |
BC129840.1 | - | 3 - 4 | 384 | 40060811 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
