Assay Details
Gene Symbol: Ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029665.3 | NP_083941.2 | 19 - 20 | 1945 | 64 | |
GenBank mRNA | AK044487.1 | - | 19 - 20 | 1977 | 64 | |
AK087793.1 | - | 19 - 20 | 1978 | 64 | ||
BC029746.1 | - | 19 - 20 | 1945 | 3494321 | 64 | |
BC031900.1 | - | 19 - 20 | 1881 | 5364208 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
