Assay Details
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176942.4 | NP_795916.1 | 4 - 5 | 619 | 81 | |
GenBank mRNA | AK038476.1 | - | 4 - 5 | 621 | 81 | |
AK083185.1 | - | 4 - 5 | 406 | 81 | ||
BC062112.1 | - | 4 - 5 | 617 | 6815819 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
