Assay Details
Gene Symbol: 9130023H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1146 | 50 | |
GenBank mRNA | AK033604.1 | - | 1 | 1106 | 50 | |
AK084086.1 | - | 1 | 1149 | 50 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
