Assay Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 8 - 9 | 715 | 87 | |
NM_001271463.1 | NP_001258392.1 | 8 - 9 | 715 | 87 | ||
NM_001271464.1 | NP_001258393.1 | 8 - 9 | 715 | 87 | ||
NM_001271465.1 | NP_001258394.1 | 8 - 9 | 742 | 87 | ||
NM_177266.5 | NP_796240.3 | 8 - 9 | 748 | 87 | ||
GenBank mRNA | AK028450.1 | - | 8 - 9 | 701 | 87 | |
AK035819.1 | - | 8 - 9 | 654 | 87 | ||
AK044371.1 | - | 8 - 9 | 716 | 87 | ||
AK083441.1 | - | 8 - 9 | 680 | 87 | ||
AK161797.1 | - | 8 - 9 | 651 | 87 | ||
AK165871.1 | - | 8 - 9 | 709 | 87 | ||
BC027341.1 | - | 8 - 9 | 699 | 3984129 | 87 | |
BC138233.1 | - | 8 - 9 | 646 | 8861252 | 87 | |
BC145208.1 | - | 8 - 9 | 646 | 9053759 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
