Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177545.5 | NP_808213.2 | 4 - 5 | 1109 | 95 | |
GenBank mRNA | AF481860.1 | - | 4 - 5 | 1025 | 95 | |
AK135230.1 | - | 4 - 5 | 1070 | 95 | ||
BC138519.1 | - | 3 - 4 | 851 | 8861539 | 95 | |
BC138520.1 | - | 3 - 4 | 851 | 8861540 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
