Assay Details
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 11 - 12 | 1378 | 109 | |
GenBank mRNA | AK086942.1 | - | 10 - 11 | 976 | 109 | |
AK132732.1 | - | 11 - 12 | 1379 | 109 | ||
AK136898.1 | - | 10 - 11 | 977 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
