Assay Details
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 7 - 8 | 970 | 89 | |
GenBank mRNA | AK086942.1 | - | 6 - 7 | 568 | 89 | |
AK132732.1 | - | 7 - 8 | 971 | 89 | ||
AK136898.1 | - | 6 - 7 | 569 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
