Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039371.2 | NP_001034460.2 | 9 - 10 | 1623 | 104 | |
GenBank mRNA | AK029695.1 | - | 6 - 7 | 781 | 104 | |
BC152347.1 | - | 4 - 5 | 518 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
