Assay Details
Gene Symbol: Cnksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177751.2 | NP_808419.1 | 14 - 15 | 1930 | 90 | |
GenBank mRNA | AK122399.1 | - | 13 - 14 | 1854 | 90 | |
BC043093.1 | - | 14 - 15 | 1930 | 6405389 | 90 | |
BC060716.1 | - | 14 - 15 | 1930 | 6405389 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
