Assay Details
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178065.4 | NP_835166.3 | 10 - 11 | 1933 | 85 | |
GenBank mRNA | AB093230.1 | - | 9 - 10 | 1552 | 85 | |
AK077015.1 | - | 9 - 10 | 1465 | 85 | ||
AK133332.1 | - | 9 - 10 | 1469 | 85 | ||
AK147491.1 | - | 10 - 11 | 1934 | 85 | ||
AK147878.1 | - | 10 - 11 | 1897 | 85 | ||
AK154529.1 | - | 10 - 11 | 1914 | 85 | ||
BC006074.1 | - | 1 - 2 | 88 | 85 | ||
BC049900.1 | - | 4 - 5 | 623 | 85 | ||
BC057351.1 | - | 10 - 11 | 1871 | 85 | ||
BC079590.1 | - | 10 - 11 | 1891 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
