Assay Details
Gene Symbol: Gpr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178760.4 | NP_848875.2 | 16 - 17 | 1477 | 88 | |
GenBank mRNA | AK030433.1 | - | 16 - 17 | 1473 | 88 | |
AK034815.1 | - | 16 - 17 | 1460 | 88 | ||
AK082237.1 | - | 16 - 17 | 1475 | 88 | ||
AK083025.1 | - | 16 - 17 | 1471 | 88 | ||
AK088544.1 | - | 16 - 17 | 1461 | 88 | ||
AK129407.1 | - | 16 - 17 | 1372 | 88 | ||
AK154530.1 | - | 16 - 17 | 1473 | 88 | ||
BC092231.1 | - | 16 - 17 | 1445 | 30607603 | 88 | |
BC117900.1 | - | 16 - 17 | 1455 | 40097045 | 88 | |
BC117901.1 | - | 16 - 17 | 1455 | 40097052 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
