Assay Details
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008740.4 | NP_032766.2 | 14 - 15 | 1703 | 66 | |
GenBank mRNA | AK028415.1 | - | 3 - 4 | 427 | 66 | |
AK032120.1 | - | 1 - 2 | 135 | 66 | ||
AK049281.1 | - | 14 - 15 | 1673 | 66 | ||
AK085086.1 | - | 14 - 15 | 1657 | 66 | ||
AK153905.1 | - | 14 - 15 | 1672 | 66 | ||
BC006627.1 | - | 14 - 15 | 1708 | 3968733 | 66 | |
BC019167.1 | - | 14 - 15 | 1656 | 4506351 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
