Assay Details
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009397.3 | NP_033423.3 | 1 - 2 | 260 | 82 | |
GenBank mRNA | AK088297.1 | - | 1 - 2 | 217 | 82 | |
AK151921.1 | - | 1 - 2 | 221 | 82 | ||
AK153238.1 | - | 1 - 2 | 215 | 82 | ||
BC050259.1 | - | 1 - 2 | 226 | 82 | ||
BC054806.1 | - | 1 - 2 | 260 | 6838303 | 82 | |
BC060221.1 | - | 1 - 2 | 260 | 6843338 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK088297.1, AK151921.1, AK153238.1, BC054806.1, BC060221.1, NM_009397.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
