Assay Details
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009602.4 | NP_033732.2 | 1 - 2 | 293 | 59 | |
GenBank mRNA | AF299083.1 | - | 1 - 2 | 87 | 59 | |
AF325349.1 | - | 1 - 2 | 53 | 59 | ||
AK049722.1 | - | 1 - 2 | 266 | 59 | ||
AK051742.1 | - | 1 - 2 | 275 | 59 | ||
AK081254.1 | - | 270 | 59 | |||
AY574267.1 | - | 1 - 2 | 67 | 59 | ||
BC065103.1 | - | 1 - 2 | 246 | 6838257 | 59 | |
BY123599.1 | - | 1 - 2 | 293 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
