Assay Details
Gene Symbol: Serinc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012032.4 | NP_036162.3 | 7 - 8 | 980 | 60 | |
GenBank mRNA | AB029499.1 | - | 7 - 8 | 889 | 60 | |
AK031101.1 | - | 7 - 8 | 969 | 60 | ||
AK128955.1 | - | 5 - 6 | 517 | 60 | ||
AK145604.1 | - | 7 - 8 | 928 | 60 | ||
AK150106.1 | - | 7 - 8 | 926 | 60 | ||
AK152720.1 | - | 7 - 8 | 926 | 60 | ||
AK155921.1 | - | 7 - 8 | 926 | 60 | ||
AK159347.1 | - | 7 - 8 | 926 | 60 | ||
AK159488.1 | - | 7 - 8 | 926 | 60 | ||
AK159819.1 | - | 7 - 8 | 926 | 60 | ||
AK160005.1 | - | 7 - 8 | 926 | 60 | ||
AK160055.1 | - | 7 - 8 | 926 | 60 | ||
AK161981.1 | - | 7 - 8 | 926 | 60 | ||
AK164807.1 | - | 7 - 8 | 926 | 60 | ||
AK166924.1 | - | 7 - 8 | 926 | 60 | ||
AK170350.1 | - | 7 - 8 | 919 | 60 | ||
BC011295.1 | - | 7 - 8 | 936 | 3487943 | 60 | |
BC022901.1 | - | 7 - 8 | 917 | 5068196 | 60 | |
BC029026.1 | - | 7 - 8 | 945 | 5039323 | 60 | |
BC029980.1 | - | 7 - 8 | 949 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
