Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031194.5 | NP_112471.3 | 1 - 2 | 204 | 73 | |
GenBank mRNA | AK031962.1 | - | 1 - 2 | 170 | 73 | |
BB622154.1 | - | 1 - 2 | 167 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK031962.1, NM_031194.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
