Assay Details
Gene Symbol: Nsun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145354.5 | NP_663329.3 | 4 - 5 | 540 | 109 | |
GenBank mRNA | AK030124.1 | - | 3 - 4 | 425 | 109 | |
AK150631.1 | - | 4 - 5 | 494 | 109 | ||
AK151917.1 | - | 4 - 5 | 494 | 109 | ||
AK154789.1 | - | 4 - 5 | 531 | 109 | ||
AK171399.1 | - | 4 - 5 | 496 | 109 | ||
BC025549.1 | - | 4 - 5 | 504 | 109 | ||
CJ092700.1 | - | 4 - 5 | 539 | 109 | ||
DQ490066.1 | - | 4 - 5 | 503 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
