Assay Details
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 4 - 5 | 413 | 61 | |
NM_146157.4 | NP_666269.2 | 4 - 5 | 413 | 61 | ||
GenBank mRNA | AK049061.1 | - | 4 - 5 | 404 | 61 | |
AK077496.1 | - | 4 - 5 | 403 | 61 | ||
AK089289.1 | - | 4 - 5 | 392 | 61 | ||
AK129052.1 | - | 4 - 5 | 366 | 61 | ||
AK138637.1 | - | 4 - 5 | 385 | 61 | ||
AK161677.1 | - | 4 - 5 | 376 | 61 | ||
AK170739.1 | - | 4 - 5 | 392 | 61 | ||
BC048948.1 | - | 13 - 14 | 61 | |||
BC129804.1 | - | 4 - 5 | 400 | 40087024 | 61 | |
BC139366.1 | - | 4 - 5 | 399 | 8862388 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
