Assay Details
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146251.4 | NP_666363.3 | 18 - 19 | 2253 | 121 | |
GenBank mRNA | AK038505.1 | - | 18 - 19 | 2139 | 121 | |
AK088362.1 | - | 18 - 19 | 2199 | 121 | ||
AK148380.1 | - | 18 - 19 | 2227 | 121 | ||
AK170417.1 | - | 18 - 19 | 2199 | 121 | ||
AK203752.1 | - | 1 - 2 | 136 | 121 | ||
BC021312.1 | - | 17 - 18 | 2287 | 121 | ||
BC025621.1 | - | 14 - 15 | 1666 | 121 | ||
BC141365.1 | - | 18 - 19 | 2037 | 9055978 | 121 | |
BC141366.1 | - | 18 - 19 | 2037 | 9055979 | 121 | |
EU293208.1 | - | 17 - 18 | 1924 | 121 | ||
EU368675.1 | - | 17 - 18 | 1924 | 121 | ||
EU402948.2 | - | 17 - 18 | 1924 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
