Assay Details
Gene Symbol: Podxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176973.4 | NP_795947.3 | 3 - 4 | 1107 | 69 | |
GenBank mRNA | AK051761.1 | - | 3 - 4 | 1310 | 69 | |
AK190030.1 | - | 1 - 2 | 362 | 69 | ||
BC033384.1 | - | 3 - 4 | 1107 | 5362581 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
