Assay Details
Gene Symbol: Slc26a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177243.4 | NP_796217.2 | 3 - 4 | 377 | 64 | |
GenBank mRNA | AK078966.1 | - | 3 - 4 | 378 | 64 | |
AK086815.1 | - | 3 - 4 | 378 | 64 | ||
AY034145.1 | - | 2 - 3 | 290 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
