Assay Details
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026560.4 | NP_080836.3 | 1 - 2 | 332 | 64 | |
GenBank mRNA | AK020070.1 | - | 1 - 2 | 288 | 64 | |
AK031571.1 | - | 1 - 2 | 260 | 64 | ||
AK076065.1 | - | 1 - 2 | 289 | 64 | ||
AK076422.1 | - | 1 - 2 | 322 | 64 | ||
AK077717.1 | - | 1 - 2 | 285 | 64 | ||
AK077920.1 | - | 1 - 2 | 255 | 64 | ||
AK078404.1 | - | 1 - 2 | 285 | 64 | ||
AK082800.1 | - | 1 - 2 | 297 | 64 | ||
AY550907.1 | - | 1 - 2 | 322 | 64 | ||
BC068181.1 | - | 1 - 2 | 280 | 30634239 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
