Assay Details
Gene Symbol: Dhx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026987.2 | NP_081263.2 | 15 - 16 | 2570 | 55 | |
GenBank mRNA | AK010431.1 | - | 2 - 3 | 188 | 55 | |
AK122314.2 | - | 15 - 16 | 2505 | 55 | ||
BC009147.1 | - | 15 - 16 | 2529 | 2647184 | 55 | |
BC046781.1 | - | 15 - 16 | 2498 | 5716182 | 55 | |
BC138204.1 | - | 15 - 16 | 2509 | 8861223 | 55 | |
BC138205.1 | - | 15 - 16 | 2509 | 8861224 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
