Assay Details
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027008.2 | NP_081284.2 | 4 - 5 | 607 | 75 | |
GenBank mRNA | AK011637.1 | - | 4 - 5 | 342 | 75 | |
AK052960.1 | - | 4 - 5 | 609 | 76 | ||
AK129077.1 | - | 4 - 5 | 567 | 75 | ||
AK143644.1 | - | 4 - 5 | 575 | 75 | ||
BC021777.1 | - | 4 - 5 | 559 | 4215964 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
