Assay Details
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027256.2 | NP_081532.1 | 16 - 17 | 2163 | 67 | |
GenBank mRNA | AK011676.1 | - | 2 - 3 | 160 | 67 | |
BC013710.1 | - | 16 - 17 | 2109 | 3156446 | 67 | |
BC013813.1 | - | 4 - 5 | 417 | 67 | ||
BC021838.1 | - | 9 - 10 | 1166 | 67 | ||
BC031145.1 | - | 10 - 11 | 1242 | 67 | ||
CX218921.1 | - | 5 - 6 | 525 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
