Assay Details
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027256.2 | NP_081532.1 | 18 - 19 | 2377 | 107 | |
GenBank mRNA | AK011676.1 | - | 4 - 5 | 374 | 107 | |
BC013710.1 | - | 18 - 19 | 2323 | 3156446 | 107 | |
BC013813.1 | - | 6 - 7 | 631 | 107 | ||
BC021838.1 | - | 11 - 12 | 1380 | 107 | ||
BC031145.1 | - | 12 - 13 | 1456 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
