Assay Details
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172073.1 | NP_001165544.1 | 7 - 8 | 979 | 80 | |
NM_028209.2 | NP_082485.1 | 7 - 8 | 978 | 80 | ||
GenBank mRNA | AK005923.1 | - | 7 - 8 | 979 | 80 | |
AK088684.1 | - | 7 - 8 | 940 | 80 | ||
AK133931.1 | - | 7 - 8 | 943 | 80 | ||
BC025435.1 | - | 7 - 8 | 927 | 5004463 | 80 | |
BC125257.1 | - | 2 - 3 | 316 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
