Assay Details
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081309.1 | NP_001074778.1 | 2 - 3 | 1171 | 90 | |
GenBank mRNA | AK042361.1 | - | 2 - 3 | 966 | 90 | |
AK042953.1 | - | 2 - 3 | 1159 | 90 | ||
AK050333.1 | - | 2 - 3 | 1141 | 90 | ||
AK054559.1 | - | 2 - 3 | 1188 | 90 | ||
AK087938.1 | - | 2 - 3 | 1162 | 90 | ||
AK169508.1 | - | 1 - 2 | 1958 | 90 | ||
BC017537.1 | - | 1 - 2 | 773 | 90 | ||
BC157947.1 | - | 2 - 3 | 996 | 9007281 | 90 | |
BC158047.1 | - | 2 - 3 | 996 | 9088160 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
