Assay Details
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030109.2 | NP_084385.2 | 5 - 6 | 683 | 72 | |
GenBank mRNA | AK012009.1 | - | 5 - 6 | 637 | 72 | |
AK012643.1 | - | 5 - 6 | 652 | 72 | ||
AK142136.1 | - | 5 - 6 | 653 | 72 | ||
AK146063.1 | - | 5 - 6 | 653 | 72 | ||
AK146540.1 | - | 5 - 6 | 648 | 72 | ||
AK146864.1 | - | 5 - 6 | 651 | 72 | ||
AK147687.1 | - | 5 - 6 | 651 | 72 | ||
AK150961.1 | - | 5 - 6 | 640 | 72 | ||
AK151354.1 | - | 5 - 6 | 630 | 72 | ||
AK152327.1 | - | 5 - 6 | 648 | 72 | ||
AK152965.1 | - | 5 - 6 | 73 | |||
AK159893.1 | - | 5 - 6 | 640 | 72 | ||
BC029745.1 | - | 5 - 6 | 618 | 72 | ||
BC045207.1 | - | 5 - 6 | 652 | 72 | ||
BC049118.1 | - | 5 - 6 | 676 | 6812422 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
