Assay Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037130.1 | NP_001032207.1 | 5 - 6 | 777 | 63 | |
NM_001165996.1 | NP_001159468.1 | 5 - 6 | 777 | 63 | ||
NM_010944.2 | NP_035074.2 | 5 - 6 | 777 | 63 | ||
GenBank mRNA | AB691772.1 | - | 5 - 6 | 627 | 63 | |
AY360453.1 | - | 5 - 6 | 672 | 63 | ||
AY956403.1 | - | 5 - 6 | 627 | 63 | ||
AY956404.1 | - | 5 - 6 | 627 | 63 | ||
U37708.1 | - | 5 - 6 | 775 | 63 | ||
U37709.1 | - | 5 - 6 | 775 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
