Assay Details
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113389.1 | NP_001106860.1 | 2 - 3 | 150 | 108 | |
NM_007890.2 | NP_031916.1 | 2 - 3 | 409 | 108 | ||
GenBank mRNA | AK140323.1 | - | 2 - 3 | 606 | 108 | |
CB526389.1 | - | 2 - 3 | 150 | 108 | ||
CD540384.1 | - | 2 - 3 | 125 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001113389.1, NM_007890.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
