Assay Details
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010578.2 | NP_034708.1 | 3 - 4 | 452 | 81 | |
GenBank mRNA | AK088016.1 | - | 3 - 4 | 353 | 81 | |
AK088729.1 | - | 3 - 4 | 199 | 81 | ||
AK159689.1 | - | 3 - 4 | 294 | 81 | ||
AK167682.1 | - | 3 - 4 | 350 | 81 | ||
BC050906.1 | - | 3 - 4 | 325 | 5721348 | 81 | |
X15202.1 | - | 2 - 3 | 145 | 81 | ||
Y00769.1 | - | 3 - 4 | 240 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
