Assay Details
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025949.3 | NP_080225.2 | 20 - 21 | 2122 | 129 | |
GenBank mRNA | AK012150.2 | - | 9 - 10 | 872 | 129 | |
AK049349.1 | - | 18 - 19 | 1873 | 129 | ||
BC054113.1 | - | 18 - 19 | 1795 | 30015425 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
