Assay Details
Gene Symbol: Gpr108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF376726.1 | - | 2 - 3 | 241 | 60 | |
AK047364.1 | - | 2 - 3 | 217 | 60 | ||
AK082797.1 | - | 2 - 3 | 246 | 60 | ||
AK159313.1 | - | 2 - 3 | 265 | 60 | ||
AK170948.1 | - | 2 - 3 | 269 | 60 | ||
AK171030.1 | - | 2 - 3 | 269 | 60 | ||
AX775895.1 | - | 2 - 3 | 277 | 60 | ||
AX775897.1 | - | 2 - 3 | 241 | 60 | ||
BB611826.1 | - | 2 - 3 | 295 | 60 | ||
BC016104.1 | - | 2 - 3 | 246 | 4920840 | 60 | |
BG079632.2 | - | 2 - 3 | 281 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
