Assay Details
Gene Symbol: Antxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054041.2 | NP_473382.1 | 12 - 13 | 1282 | 72 | |
GenBank mRNA | AF378762.1 | - | 12 - 13 | 1222 | 72 | |
AK013005.1 | - | 10 - 11 | 689 | 72 | ||
AK031465.1 | - | 12 - 13 | 1272 | 72 | ||
BC057043.1 | - | 11 - 12 | 746 | 72 | ||
BC094544.1 | - | 12 - 13 | 1227 | 6406827 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
