Assay Details
Gene Symbol: Iws1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173441.3 | NP_775617.1 | 1 - 2 | 300 | 73 | |
GenBank mRNA | AK028247.1 | - | 1 - 2 | 271 | 73 | |
AK033013.1 | - | 1 - 2 | 295 | 73 | ||
AK041691.1 | - | 1 - 2 | 294 | 73 | ||
AK163972.1 | - | 1 - 2 | 271 | 73 | ||
BC068184.1 | - | 1 - 2 | 240 | 30639737 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK028247.1, AK041691.1, AK163972.1, BC068184.1, NM_173441.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
