Assay Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175749.2 | NP_786925.2 | 2 - 3 | 715 | 98 | |
GenBank mRNA | AK086847.1 | - | 2 - 3 | 716 | 98 | |
BC052173.1 | - | 2 - 3 | 453 | 6516328 | 98 | |
CF898039.1 | - | 2 - 3 | 353 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
