Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700032239.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032239.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032239.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032244.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032244.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032244.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032229.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032229.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032229.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032238.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032238.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032238.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032250.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032250.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032250.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032232.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032232.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032232.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032234.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032234.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032234.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032231.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032231.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032231.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032233.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032233.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032233.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032227.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032227.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032227.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032230.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032230.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032230.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032235.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032235.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032235.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032251.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032251.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032251.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032228.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032228.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032228.mm.1 |
TC0700003826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032242.mm.1 |
NM_177073.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032242.mm.1 |
NM_177073.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032242.mm.1 |
TC0700003826.mm.1 |
MTA TC |