Assay Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145952.1 | NP_001139424.1 | 5 - 6 | 647 | 86 | |
NM_178665.5 | NP_848780.3 | 6 - 7 | 854 | 86 | ||
GenBank mRNA | AK029335.1 | - | 6 - 7 | 646 | 86 | |
AK040185.1 | - | 1 - 2 | 170 | 86 | ||
AK040643.1 | - | 4 - 5 | 408 | 86 | ||
AK045288.1 | - | 6 - 7 | 715 | 86 | ||
AK045341.1 | - | 6 - 7 | 698 | 86 | ||
AK051345.1 | - | 6 - 7 | 726 | 86 | ||
AK054550.1 | - | 5 - 6 | 650 | 86 | ||
AK085626.1 | - | 6 - 7 | 846 | 86 | ||
AK086307.1 | - | 6 - 7 | 665 | 86 | ||
AK168431.1 | - | 6 - 7 | 760 | 86 | ||
AK169780.1 | - | 6 - 7 | 666 | 86 | ||
CG784727.1 | - | 2 - 3 | 238 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
