Assay Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145952.1 | NP_001139424.1 | 6 - 7 | 776 | 68 | |
NM_178665.5 | NP_848780.3 | 7 - 8 | 983 | 68 | ||
GenBank mRNA | AK040643.1 | - | 5 - 6 | 537 | 68 | |
AK045288.1 | - | 7 - 8 | 844 | 68 | ||
AK051345.1 | - | 7 - 8 | 855 | 68 | ||
AK054550.1 | - | 6 - 7 | 779 | 68 | ||
AK085626.1 | - | 7 - 8 | 975 | 68 | ||
AK086307.1 | - | 7 - 8 | 794 | 68 | ||
AK169780.1 | - | 7 - 8 | 795 | 68 | ||
BC085321.1 | - | 6 - 7 | 722 | 30743832 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
