Assay Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145952.1 | NP_001139424.1 | 8 - 9 | 1588 | 77 | |
NM_001145954.1 | NP_001139426.1 | 4 - 5 | 1002 | 77 | ||
NM_178665.5 | NP_848780.3 | 9 - 10 | 1795 | 77 | ||
GenBank mRNA | AK029567.1 | - | 4 - 5 | 1003 | 77 | |
AK040643.1 | - | 7 - 8 | 1349 | 77 | ||
AK045288.1 | - | 9 - 10 | 1656 | 77 | ||
AK051345.1 | - | 9 - 10 | 1667 | 77 | ||
AK054550.1 | - | 8 - 9 | 1591 | 77 | ||
AK076989.1 | - | 4 - 5 | 993 | 77 | ||
AK085626.1 | - | 9 - 10 | 1788 | 77 | ||
AK168431.1 | - | 9 - 10 | 1702 | 77 | ||
AK169780.1 | - | 9 - 10 | 1607 | 77 | ||
BC085321.1 | - | 8 - 9 | 1534 | 30743832 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Smooth muscle expression of lipoma preferred partner is mediated by an alternative intronic promoter that is regulated by serum response factor/myocardin.
- Authors
- Petit MM, Lindskog H, Larsson E, Wasteson P, Athley E, Breuer S, Angstenberger M, Hertfelder D, Mattsson E, Nordheim A, Nelander S, Lindahl P
- Journal
- Circ Res - Page No 61 (Thu Jul 03 00:00:00 UTC 2008)
- Volume (Year)
- 103 (2008)
- Product Usage
- -
