Assay Details
Gene Symbol: Clca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178697.4 | NP_848812.1 | 5 - 6 | 816 | 92 | |
GenBank mRNA | AK028704.1 | - | 5 - 6 | 817 | 92 | |
AK035512.1 | - | 5 - 6 | 883 | 92 | ||
AK048276.1 | - | 5 - 6 | 807 | 92 | ||
AY161007.1 | - | 5 - 6 | 772 | 92 | ||
BC096379.1 | - | 5 - 6 | 879 | 5317084 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
