Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 5 - 6 | 868 | 100 | |
GenBank mRNA | AK142280.1 | - | 5 - 6 | 860 | 100 | |
AK150089.1 | - | 5 - 6 | 860 | 100 | ||
AK157252.1 | - | 5 - 6 | 830 | 100 | ||
AK160880.1 | - | 5 - 6 | 860 | 100 | ||
AK160890.1 | - | 5 - 6 | 846 | 100 | ||
AK167582.1 | - | 5 - 6 | 856 | 100 | ||
BC027747.1 | - | 5 - 6 | 857 | 4459657 | 100 | |
BC068193.1 | - | 5 - 6 | 827 | 6813997 | 100 | |
X92411.1 | - | 5 - 6 | 548 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
