Assay Details
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007386.2 | NP_031412.2 | 6 - 7 | 964 | 59 | |
GenBank mRNA | AK027914.1 | - | 6 - 7 | 750 | 59 | |
AK085951.1 | - | 6 - 7 | 713 | 59 | ||
AK129025.1 | - | 6 - 7 | 746 | 59 | ||
AK163985.1 | - | 6 - 7 | 763 | 59 | ||
AK194174.1 | - | 1 - 2 | 115 | 59 | ||
AK201556.1 | - | 1 - 2 | 116 | 59 | ||
BC005454.1 | - | 6 - 7 | 724 | 3494686 | 59 | |
X61147.1 | - | 6 - 7 | 731 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
