Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 6 - 7 | 732 | 71 | |
NM_009422.3 | NP_033448.2 | 6 - 7 | 711 | 71 | ||
GenBank mRNA | AF027570.1 | - | 5 - 6 | 623 | 71 | |
AK038136.1 | - | 6 - 7 | 660 | 71 | ||
AK052934.1 | - | 6 - 7 | 660 | 71 | ||
AK151653.1 | - | 6 - 7 | 649 | 71 | ||
AK152174.1 | - | 6 - 7 | 679 | 71 | ||
AK159581.1 | - | 6 - 7 | 681 | 71 | ||
AK159585.1 | - | 658 | 71 | |||
BC003801.1 | - | 5 - 6 | 637 | 3492839 | 71 | |
BC060625.1 | - | 4 - 5 | 2668 | 71 | ||
L35303.1 | - | 6 - 7 | 648 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
